Anti-MSI2 Mouse Monoclonal Antibody [clone: 3C6]

Lieferant: Abnova
H00124540-M09
ABNOH00124540-M09EA 397 EUR
ABNOH00124540-M09
Anti-MSI2 Mouse Monoclonal Antibody [clone: 3C6]
Antikörper
Type: Primary
Antigen: MSI2
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR