Anti-SEMA3D Rabbit Polyclonal Antibody

Lieferant: Abnova
H00223117-D01P
ABNOH00223117-D01PEA 364 EUR
ABNOH00223117-D01P
Anti-SEMA3D Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human SEMA3D protein.

Recommended Dilutons: Western Blot (1 µg/ml).

Type: Primary
Antigen: SEMA3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR