Anti-NDUFS7 Mouse Monoclonal Antibody [clone: 3A3]

Lieferant: Abnova
H00374291-M01
ABNOH00374291-M01EA 397 EUR
ABNOH00374291-M01
Anti-NDUFS7 Mouse Monoclonal Antibody [clone: 3A3]
Antikörper
Type: Primary
Antigen: NDUFS7
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR