Anti-SUMO4 Mouse Monoclonal Antibody [clone: 1D3]

Lieferant: Abnova
H00387082-M02
ABNOH00387082-M02EA 397 EUR
ABNOH00387082-M02
Anti-SUMO4 Mouse Monoclonal Antibody [clone: 1D3]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant SUMO4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SUMO4
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR