Anti-APMAP Mouse Monoclonal Antibody [clone: 4F5]

Lieferant: Abnova
MAB0037-M04
ABNOMAB0037-M04EA 409 EUR
ABNOMAB0037-M04
Anti-APMAP Mouse Monoclonal Antibody [clone: 4F5]
Antikörper
Mouse monoclonal antibody raised against adiponectin.

Type: Primary
Antigen: APMAP
Clonality: Monoclonal
Clone: 4F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR