Anti-SLC17A6 Mouse Monoclonal Antibody (ATTO 594) [clone: S29-29]

Lieferant: Abnova
MAB11534
ABNOMAB11534EA 912 EUR
ABNOMAB11534
Anti-SLC17A6 Mouse Monoclonal Antibody (ATTO 594) [clone: S29-29]
Antikörper
Mouse monoclonal antibody raised against recombinant of Slc17a6.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A6
Clonality: Monoclonal
Clone: S29-29
Conjugation: ATTO 594
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR