Anti-CACNA1G Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S178A-9]

Lieferant: Abnova
MAB11557
ABNOMAB11557EA 648 EUR
ABNOMAB11557
Anti-CACNA1G Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S178A-9]
Antikörper
Mouse monoclonal antibody raised against recombinant of Cacna1g.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR