Anti-PMEL Mouse Monoclonal Antibody [clone: SPM142]

Lieferant: Abnova
MAB12053
ABNOMAB12053EA 523 EUR
ABNOMAB12053
Anti-PMEL Mouse Monoclonal Antibody [clone: SPM142]
Antikörper
Mouse monoclonal antibody raised against SILV.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PMEL
Clonality: Monoclonal
Clone: SPM142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR