Anti-SPN Mouse Monoclonal Antibody [clone: SPM503]

Lieferant: Abnova
MAB12056
ABNOMAB12056EA 523 EUR
ABNOMAB12056
Anti-SPN Mouse Monoclonal Antibody [clone: SPM503]
Antikörper
Mouse monoclonal antibody raised against SPN.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/106 cells; Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: SPM503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR