Anti-NKX2-2 Mouse Monoclonal Antibody [clone: SPM564]

Lieferant: Abnova
MAB12120
ABNOMAB12120EA 523 EUR
ABNOMAB12120
Anti-NKX2-2 Mouse Monoclonal Antibody [clone: SPM564]
Antikörper
Mouse monoclonal antibody raised against NKX2-2.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: SPM564
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR