Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]

Lieferant: Abnova
MAB12122
ABNOMAB12122EA 523 EUR
ABNOMAB12122
Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]
Antikörper
Mouse monoclonal antibody raised against PCNA.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: SPM350
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR