Anti-CD68 Mouse Monoclonal Antibody [clone: C68/684]

Lieferant: Abnova
MAB12126
ABNOMAB12126EA 523 EUR
ABNOMAB12126
Anti-CD68 Mouse Monoclonal Antibody [clone: C68/684]
Antikörper
Mouse monoclonal antibody raised against human CD68.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: C68/684
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR