Anti-IGKC Mouse Monoclonal Antibody [clone: KLC709]

Lieferant: Abnova
MAB12132
ABNOMAB12132EA 523 EUR
ABNOMAB12132
Anti-IGKC Mouse Monoclonal Antibody [clone: KLC709]
Antikörper
Mouse monoclonal antibody raised against recombinant human IGKC.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: KLC709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR