Anti-DSG1 Mouse Monoclonal Antibody [clone: CC49]

Lieferant: Abnova
MAB12146
ABNOMAB12146EA 523 EUR
ABNOMAB12146
Anti-DSG1 Mouse Monoclonal Antibody [clone: CC49]
Antikörper
Mouse monoclonal antibody raised against human DSG1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone: CC49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR