Anti-CEA260 Mouse Monoclonal Antibody [clone: C66/1260]

Lieferant: Abnova
MAB13253
ABNOMAB13253EA 523 EUR
ABNOMAB13253
Anti-CEA260 Mouse Monoclonal Antibody [clone: C66/1260]
Antikörper
Mouse monoclonal antibody raised against native human Carcinoembryonic Antigen (CEA).

Type: Primary
Antigen: CEA260
Clonality: Monoclonal
Clone: C66/1260
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR