Anti-CEA009 Mouse Monoclonal Antibody [clone: C66/1009]

Lieferant: Abnova
MAB13258
ABNOMAB13258EA 523 EUR
ABNOMAB13258
Anti-CEA009 Mouse Monoclonal Antibody [clone: C66/1009]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human Carcinoembryonic Antigen (CEA).

Type: Primary
Antigen: CEA009
Clonality: Monoclonal
Clone: C66/1009
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR