Anti-ERBB2776 Mouse Monoclonal Antibody [clone: ERB2/776]

Lieferant: Abnova
MAB13267
ABNOMAB13267EA 523 EUR
ABNOMAB13267
Anti-ERBB2776 Mouse Monoclonal Antibody [clone: ERB2/776]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human ERBB2.

Type: Primary
Antigen: ERBB2776
Clonality: Monoclonal
Clone: ERB2/776
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR