Anti-GFAP/789 Mouse Monoclonal Antibody [clone: ASTRO/789]

Lieferant: Abnova
MAB13278
ABNOMAB13278EA 523 EUR
ABNOMAB13278
Anti-GFAP/789 Mouse Monoclonal Antibody [clone: ASTRO/789]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human GFAP.

Type: Primary
Antigen: GFAP/789
Clonality: Monoclonal
Clone: ASTRO/789
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1
Reactivity: Human,Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR