Anti-B3GAT14 Mouse Monoclonal Antibody [clone: NK/804]

Lieferant: Abnova
MAB13284
ABNOMAB13284EA 523 EUR
ABNOMAB13284
Anti-B3GAT14 Mouse Monoclonal Antibody [clone: NK/804]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human B3GAT1.

Type: Primary
Antigen: B3GAT14
Clonality: Monoclonal
Clone: NK/804
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR