Anti-SPN39 Mouse Monoclonal Antibody [clone: SPN/839]

Lieferant: Abnova
MAB13434
ABNOMAB13434EA 523 EUR
ABNOMAB13434
Anti-SPN39 Mouse Monoclonal Antibody [clone: SPN/839]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human SPN.

Type: Primary
Antigen: SPN39
Clonality: Monoclonal
Clone: SPN/839
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR