Anti-SPN1 Mouse Monoclonal Antibody [clone: 84-3C1]

Lieferant: Abnova
MAB13435
ABNOMAB13435EA 523 EUR
ABNOMAB13435
Anti-SPN1 Mouse Monoclonal Antibody [clone: 84-3C1]
Antikörper
Mouse monoclonal antibody raised against native human SPN.

Type: Primary
Antigen: SPN1
Clonality: Monoclonal
Clone: 84-3C1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR