Anti-SPN1 Mouse Monoclonal Antibody [clone: 84-3C1]
Lieferant: Abnova
MAB13436
ABNOMAB13436EA 523 EUR
ABNOMAB13436
Anti-SPN1 Mouse Monoclonal Antibody [clone: 84-3C1]
Antikörper
Mouse monoclonal antibody raised against native human SPN.
Type: Primary
Antigen: SPN1
Clonality: Monoclonal
Clone: 84-3C1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human