Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: HM031]

Lieferant: Abnova
MAB13498
ABNOMAB13498EA 648 EUR
ABNOMAB13498
Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: HM031]
Antikörper
Mouse monoclonal antibody raised against Chlamydia trachomatis.

Type: Primary
Antigen: Chlamydia trachomatis
Clonality: Monoclonal
Clone: HM031
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chlamydia Species
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR