Anti-Feline leukemia virus p27 Mouse Monoclonal Antibody [clone: HM189]

Lieferant: Abnova
MAB13548
ABNOMAB13548EA 912 EUR
ABNOMAB13548
Anti-Feline leukemia virus p27 Mouse Monoclonal Antibody [clone: HM189]
Antikörper
Mouse monoclonal antibody raised against feline leukemia virus p27.

Type: Primary
Antigen: Feline leukemia virus p27
Clonality: Monoclonal
Clone: HM189
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Viruses
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR