Anti-Influenza A virus Mouse Monoclonal Antibody [clone: J320]

Lieferant: Abnova
MAB13601
ABNOMAB13601EA 648 EUR
ABNOMAB13601
Anti-Influenza A virus Mouse Monoclonal Antibody [clone: J320]
Antikörper
Mouse monoclonal antibody raised against influenza A virus.

Type: Primary
Antigen: Influenza A virus
Clonality: Monoclonal
Clone: J320
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Influenza virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR