Anti-Metapneumovirus Mouse Monoclonal Antibody [clone: HM043]

Lieferant: Abnova
MAB13618
ABNOMAB13618EA 648 EUR
ABNOMAB13618
Anti-Metapneumovirus Mouse Monoclonal Antibody [clone: HM043]
Antikörper
Mouse monoclonal antibody raised against metapneumovirus.

Type: Primary
Antigen: Metapneumovirus
Clonality: Monoclonal
Clone: HM043
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viruses
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR