Anti-Norovirus group II Mouse Monoclonal Antibody [clone: HM025]

Lieferant: Abnova
MAB13626
ABNOMAB13626EA 648 EUR
ABNOMAB13626
Anti-Norovirus group II Mouse Monoclonal Antibody [clone: HM025]
Antikörper
Mouse monoclonal antibody raised against norovirus group II.

Type: Primary
Antigen: Norovirus group II
Clonality: Monoclonal
Clone: HM025
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viruses
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR