Anti-LHB Mouse Monoclonal Antibody [clone: LHb/1214]

Lieferant: Abnova
MAB14354
ABNOMAB14354EA 523 EUR
ABNOMAB14354
Anti-LHB Mouse Monoclonal Antibody [clone: LHb/1214]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human LHB.

Type: Primary
Antigen: LHB
Clonality: Monoclonal
Clone: LHb/1214
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR