Anti-ODC1 Mouse Monoclonal Antibody [clone: ODC1/487]
Lieferant: Abnova
MAB14389
ABNOMAB14389EA 523 EUR
ABNOMAB14389
Anti-ODC1 Mouse Monoclonal Antibody [clone: ODC1/487]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human ODC1.
Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: ODC1/487
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Rat