Anti-Testosterone-3-carboxy-methyl-oxyme conjugated to BSA Mouse Monoclonal Antibody [clone: 4E1G2]

Lieferant: Abnova
MAB14425
ABNOMAB14425EA 523 EUR
ABNOMAB14425
Anti-Testosterone-3-carboxy-methyl-oxyme conjugated to BSA Mouse Monoclonal Antibody [clone: 4E1G2]
Antikörper
Mouse monoclonal antibody raised against human testosterone.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR