Anti-NKX2-1 Mouse Monoclonal Antibody [clone: NX2.1/690]

Lieferant: Abnova
MAB14451
ABNOMAB14451EA 523 EUR
ABNOMAB14451
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: NX2.1/690]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human NKX2-1.

Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: NX2.1/690
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR