Anti-CLTA Mouse Monoclonal Antibody [clone: CLC/1421]

Lieferant: Abnova
MAB14481
ABNOMAB14481EA 523 EUR
ABNOMAB14481
Anti-CLTA Mouse Monoclonal Antibody [clone: CLC/1421]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human CLTA/CLTB.

Type: Primary
Antigen: CLTA
Clonality: Monoclonal
Clone: CLC/1421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR