Anti-DES Mouse Monoclonal Antibody [clone: DES/1711]
Lieferant: Abnova
MAB14497
ABNOMAB14497EA 523 EUR
ABNOMAB14497
Anti-DES Mouse Monoclonal Antibody [clone: DES/1711]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human DES.
Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: DES/1711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human