Anti-EGFR Mouse Monoclonal Antibody [clone: 31G7 + GFR1195]

Lieferant: Abnova
MAB14509
ABNOMAB14509EA 523 EUR
ABNOMAB14509
Anti-EGFR Mouse Monoclonal Antibody [clone: 31G7 + GFR1195]
Antikörper
Mouse monoclonal antibody raised against human EGFR.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 31G7 + GFR1195
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR