Anti-EGFR Mouse Monoclonal Antibody [clone: GFR/1708]
Lieferant: Abnova
MAB14511
ABNOMAB14511EA 523 EUR
ABNOMAB14511
Anti-EGFR Mouse Monoclonal Antibody [clone: GFR/1708]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human EGFR.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: GFR/1708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human