Anti-ENG Mouse Monoclonal Antibody [clone: ENG/1326]

Lieferant: Abnova
MAB14512
ABNOMAB14512EA 523 EUR
ABNOMAB14512
Anti-ENG Mouse Monoclonal Antibody [clone: ENG/1326]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human ENG.

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: ENG/1326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR