Anti-EPO Mouse Monoclonal Antibody [clone: EPO/1368]

Lieferant: Abnova
MAB14518
ABNOMAB14518EA 523 EUR
ABNOMAB14518
Anti-EPO Mouse Monoclonal Antibody [clone: EPO/1368]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human EPO.

Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone: EPO/1368
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR