Anti-IGLC1 Mouse Monoclonal Antibody [clone: LLC/1738]

Lieferant: Abnova
MAB14566
ABNOMAB14566EA 523 EUR
ABNOMAB14566
Anti-IGLC1 Mouse Monoclonal Antibody [clone: LLC/1738]
Antikörper
Mouse monoclonal antibody raised against native human Lambda Light Chain .

Type: Primary
Antigen: IGLC1
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR