Anti-Testosterone-3-carboxy-methyl-oxyme Mouse Monoclonal Antibody (BSA (Bovine Serum Albumin)) [clone: 4E1G2]

Lieferant: Abnova
MAB14711
ABNOMAB14711EA 523 EUR
ABNOMAB14711
Anti-Testosterone-3-carboxy-methyl-oxyme Mouse Monoclonal Antibody (BSA (Bovine Serum Albumin)) [clone: 4E1G2]
Antikörper
Mouse monoclonal antibody raised against human testosterone.

Type: Primary
Antigen: Testosterone-3-carboxy-methyl-oxyme
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: BSA (Bovine Serum Albumin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR