Anti-HPV-16, major capsid protein L1. Mouse Monoclonal Antibody [clone: HPV16L1/1058]

Lieferant: Abnova
MAB14716
ABNOMAB14716EA 523 EUR
ABNOMAB14716
Anti-HPV-16, major capsid protein L1. Mouse Monoclonal Antibody [clone: HPV16L1/1058]
Antikörper
Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16).

Type: Primary
Antigen: HPV-16, major capsid protein L1.
Clonality: Monoclonal
Clone: HPV16L1/1058
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR