Anti-SERPINA3 Mouse Monoclonal Antibody [clone: AACT/1452]

Lieferant: Abnova
MAB14735
ABNOMAB14735EA 523 EUR
ABNOMAB14735
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: AACT/1452]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human SERPINA3.

Type: Primary
Antigen: SERPINA3
Clonality: Monoclonal
Clone: AACT/1452
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR