Anti-CLTC Mouse Monoclonal Antibody [clone: CLTC/1431]

Lieferant: Abnova
MAB14739
ABNOMAB14739EA 523 EUR
ABNOMAB14739
Anti-CLTC Mouse Monoclonal Antibody [clone: CLTC/1431]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human CLTC.

Type: Primary
Antigen: CLTC
Clonality: Monoclonal
Clone: CLTC/1431
Conjugation:
Epitope: N-terminal
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR