Anti-ANO1 Mouse Monoclonal Antibody [clone: DG1/1485]

Lieferant: Abnova
MAB14889
ABNOMAB14889EA 523 EUR
ABNOMAB14889
Anti-ANO1 Mouse Monoclonal Antibody [clone: DG1/1485]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human ANO1.

Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: DG1/1485
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR