Anti-TOP2A Mouse Monoclonal Antibody [clone: TOP2A/1361]

Lieferant: Abnova
MAB15013
ABNOMAB15013EA 523 EUR
ABNOMAB15013
Anti-TOP2A Mouse Monoclonal Antibody [clone: TOP2A/1361]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human TOP2A.

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: TOP2A/1361
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR