Anti-TSHR Mouse Monoclonal Antibody [clone: TSHRA/1402]

Lieferant: Abnova
MAB15021
ABNOMAB15021EA 523 EUR
ABNOMAB15021
Anti-TSHR Mouse Monoclonal Antibody [clone: TSHRA/1402]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human TSHR, A Chain.

Type: Primary
Antigen: TSHR
Clonality: Monoclonal
Clone: TSHRA/1402
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR