Anti-TSHR Mouse Monoclonal Antibody [clone: SPM222]

Lieferant: Abnova
MAB15026
ABNOMAB15026EA 523 EUR
ABNOMAB15026
Anti-TSHR Mouse Monoclonal Antibody [clone: SPM222]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human TSHR, B Chain.

Type: Primary
Antigen: TSHR
Clonality: Monoclonal
Clone: SPM222
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR