Anti-VWF Mouse Monoclonal Antibody [clone: VWF/1767]

Lieferant: Abnova
MAB15039
ABNOMAB15039EA 523 EUR
ABNOMAB15039
Anti-VWF Mouse Monoclonal Antibody [clone: VWF/1767]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human VWF.

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: VWF/1767
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR