Anti-FN1 Mouse Monoclonal Antibody [clone: SPM539]
Lieferant: Abnova
MAB15190
ABNOMAB15190EA 523 EUR
ABNOMAB15190
Anti-FN1 Mouse Monoclonal Antibody [clone: SPM539]
Antikörper
Mouse monoclonal antibody raised against native human FN1.
Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: SPM539
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human