Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM569 + SPM570]
Lieferant: Abnova
MAB15209
ABNOMAB15209EA 523 EUR
ABNOMAB15209
Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM569 + SPM570]
Antikörper
Mouse monoclonal antibody raised against native human PTPRC.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: SPM569 + SPM570
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human