Anti-GSK3A Mouse Monoclonal Antibody [clone: 9D5G1]

Lieferant: Abnova
MAB15269
ABNOMAB15269EA 423 EUR
ABNOMAB15269
Anti-GSK3A Mouse Monoclonal Antibody [clone: 9D5G1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant human GSK3A.

Type: Primary
Antigen: GSK3A
Clonality: Monoclonal
Clone: 9D5G1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR